siRNA Details
Gene Symbol: Gm609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005854.2 | NP_001005854.2 | 2, 3 | 449 | ||
GenBank mRNA | AB853320.1 | - | 2, 3 | 465 | ||
AK136536.1 | - | 2, 3 | 467 | |||
BC054854.1 | - | 2, 3 | 448 | 6529072 |
