siRNA Details
Gene Symbol: Exog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172136.1 | NP_001165607.1 | 6 | 731 | ||
NM_172456.3 | NP_766044.1 | 5 | 696 | |||
GenBank mRNA | AK028887.1 | - | 5 | 660 | ||
AK034654.1 | - | 6 | 693 | |||
AK051300.1 | - | 6 | 695 | |||
AK162566.1 | - | 5 | 659 | |||
AK163555.1 | - | 5 | 660 | |||
BC052171.1 | - | 4 | 502 | |||
BC059920.1 | - | 6 | 674 | |||
BC064730.1 | - | 6 | 746 | |||
BC094589.1 | - | 3 | 3182 | |||
BC119024.1 | - | 5 | 660 | 8733773 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
