siRNA Details
Gene Symbol: Tor1aip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160018.1 | NP_001153490.1 | 10 | 1695 | ||
NM_001160019.1 | NP_001153491.1 | 10 | 1109 | |||
NM_144791.2 | NP_659040.2 | 7 | 1527 | |||
GenBank mRNA | AB251963.1 | - | 11 | 1692 | ||
AK152751.1 | - | 10 | 1129 | |||
BC010841.1 | - | 7 | 1342 | 3493883 |

More Information
Other Species Targeted

Gene Ontology Categories:
