siRNA Details
Gene Symbol: Iqcj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177585.3 | NP_808253.1 | 2 | 146 | ||
GenBank mRNA | AK052136.1 | - | 2 | 147 | ||
BC107305.1 | - | 2 | 110 | 40053239 | ||
BC107306.1 | - | 2 | 110 | 40053241 |
Gene Symbol: Gm21949 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113419.2 | NP_001106890.1 | 2 | 146 | ||
GenBank mRNA | KJ941154.1 | - | 2 | 134 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
