siRNA Details
Gene Symbol: 1810043H04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110242.1 | NP_001103712.1 | 3 | 422 | ||
GenBank mRNA | AK007756.1 | - | 3 | 420 | ||
AK010111.1 | - | 3 | 420 | |||
BC061009.1 | - | 1 | 132 | |||
BC147711.1 | - | 3 | 402 | 9087470 | ||
BC147720.1 | - | 3 | 402 | 9087479 |
