siRNA Details
Gene Symbol: 1810043H04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110242.1 | NP_001103712.1 | 3 | 461 | ||
GenBank mRNA | AK007756.1 | - | 3 | 459 | ||
BC147711.1 | - | 3 | 441 | 9087470 | ||
BC147720.1 | - | 3 | 441 | 9087479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
