siRNA Details
Gene Symbol: Tmem212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164437.1 | NP_001157909.1 | 3 | 393 | ||
GenBank mRNA | AK053134.1 | - | 3 | 393 | ||
BC119391.1 | - | 3 | 393 | 8734140 | ||
BC119393.1 | - | 3 | 393 | 8734142 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
