siRNA Details
Gene Symbol: Etl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081006.1 | NP_001074475.1 | 4 | 680 | ||
NM_001177630.2 | NP_001171101.2 | 3 | 778 | |||
NM_001177631.1 | NP_001171102.1 | 3 | 778 | |||
NM_029895.4 | NP_084171.2 | 4 | 991 | |||
NM_178059.5 | NP_835160.2 | 4 | 991 | |||
GenBank mRNA | AB093289.2 | - | 3 | 568 | ||
AB125594.2 | - | 4 | 991 | |||
AB125595.2 | - | 4 | 991 | |||
AK052020.1 | - | 5 | 1093 | |||
AK052733.1 | - | 3 | 466 | |||
AK147510.1 | - | 3 | 685 | |||
BC050016.1 | - | 3 | 481 | |||
BC158051.1 | - | 3 | 500 | 9007405 | ||
BC172143.1 | - | 3 | 500 | |||
CF744489.1 | - | 4 | 680 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
