siRNA Details
Gene Symbol: Eif5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177586.5 | NP_808254.1 | 3 | 377 | ||
GenBank mRNA | AK036127.1 | - | 3 | 379 | ||
AK052412.1 | - | 3 | 321 | |||
AK082214.1 | - | 3 | 294 | |||
AY205262.1 | - | 3 | 377 | |||
AY205263.1 | - | 3 | 377 | |||
AY205264.1 | - | 3 | 377 | |||
AY205265.1 | - | 3 | 377 | |||
BC106843.2 | - | 3 | 307 | 40045849 | ||
BC106844.1 | - | 3 | 307 | 40045850 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
