siRNA Details
Gene Symbol: Daam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286452.1 | NP_001273381.1 | 20 | 2827 | ||
NM_026102.3 | NP_080378.2 | 21 | 2822 | |||
NM_172464.3 | NP_766052.2 | 21 | 2687 | |||
GenBank mRNA | AK129185.1 | - | 21 | 2665 | ||
AK147480.1 | - | 20 | 2641 | |||
AY426535.1 | - | 21 | 2732 | |||
BC025638.1 | - | 19 | 2557 | |||
BC032287.1 | - | 7 | 731 | |||
BC038403.1 | - | 20 | 2601 | |||
BC047431.1 | - | 21 | 2609 | |||
BC048856.1 | - | 1 | 150 | |||
BC076585.1 | - | 21 | 2677 | 30622306 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
