siRNA Details
Gene Symbol: Gm595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085499.1 | NP_001078968.1 | 9 | 1145 | ||
GenBank mRNA | AK133392.1 | - | 9 | 1147 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085499.1 | NP_001078968.1 | 9 | 1145 | ||
GenBank mRNA | AK133392.1 | - | 9 | 1147 |