siRNA Details
Gene Symbol: Sirt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153056.2 | NP_694696.2 | 7, 8 | 847 | ||
GenBank mRNA | AB221687.1 | - | 3, 4 | 367 | ||
AY251540.1 | - | 7, 8 | 823 | |||
BC026403.1 | - | 7, 8 | 812 | 4209498 | ||
BC026637.1 | - | 6, 7 | 888 | |||
BC026650.1 | - | 7, 8 | 815 | 4987746 | ||
BC040759.1 | - | 7, 8 | 812 | 4209498 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
