siRNA Details
Gene Symbol: Vps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285893.1 | NP_001272822.1 | 40 | 3779 | ||
NM_001285894.1 | NP_001272823.1 | 40 | 3642 | |||
GenBank mRNA | AK029007.1 | - | 24 | 1996 | ||
AK031205.1 | - | 17 | 1821 | |||
AK122375.1 | - | 40 | 3591 | |||
AK141678.1 | - | 40 | 3702 | |||
BC023914.1 | - | 26 | 2170 | 5323126 | ||
BC032214.1 | - | 20 | 1748 | 5323693 | ||
BC055323.1 | - | 40 | 3696 | 5706288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
