siRNA Details
Gene Symbol: Zc3hav1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172467.3 | NP_766055.1 | 3 | 769 | ||
GenBank mRNA | AK045287.1 | - | 3 | 770 | ||
AK051064.1 | - | 3 | 771 | |||
AK051832.1 | - | 1 | 158 | |||
AK084475.1 | - | 3 | 701 | |||
BC138180.1 | - | 3 | 861 | 8861199 | ||
BC138181.1 | - | 3 | 861 | 8861200 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
