siRNA Details
Gene Symbol: Ccnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183015.3 | NP_898836.2 | 7 | 2903 | ||
GenBank mRNA | AJ555464.1 | - | 7 | 2904 | ||
BC132411.1 | - | 6 | 2857 | 40130688 | ||
BC138355.1 | - | 6 | 2857 | 8861375 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
