siRNA Details
Gene Symbol: Snx30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172468.2 | NP_766056.1 | 5 | 1165 | ||
GenBank mRNA | AK029012.1 | - | 5 | 967 | ||
AK076374.1 | - | 2 | 169 | |||
AK147437.1 | - | 5 | 1167 | |||
AK165619.1 | - | 5 | 975 | |||
AK170774.1 | - | 5 | 939 | |||
AK170999.1 | - | 5 | 967 | |||
BC099674.1 | - | 5 | 961 | 4950374 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
