siRNA Details
Gene Symbol: Ido2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145949.2 | NP_666061.3 | 4, 5 | 523 | ||
GenBank mRNA | AK082385.1 | - | 3, 4 | 714 | ||
BC026393.1 | - | 4, 5 | 523 | 4195134 | ||
EF137182.1 | - | 4, 5 | 523 | |||
EU440733.1 | - | 3, 4 | 315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
