siRNA Details
Gene Symbol: Acnat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145368.2 | NP_663343.2 | 5 | 1117 | ||
GenBank mRNA | AK050308.1 | - | 5 | 1117 | ||
AK050357.1 | - | 6 | 1114 | |||
BC010829.1 | - | 3 | 961 | 4220620 | ||
EU564338.1 | - | 3 | 990 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
