siRNA Details
Gene Symbol: Csta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033239.3 | NP_001028411.1 | 3 | 360 | ||
GenBank mRNA | AK137702.1 | - | 3 | 361 | ||
BC139156.1 | - | 3 | 255 | 8862178 | ||
BC139157.1 | - | 3 | 255 | 8862179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
