siRNA Details
Gene Symbol: Hace1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172473.3 | NP_766061.2 | 24 | 2894 | ||
GenBank mRNA | AK029162.1 | - | 14 | 1714 | ||
AK039378.1 | - | 2 | 377 | |||
AK042879.1 | - | 24 | 2835 | |||
AK156958.1 | - | 24 | 2836 | |||
AK220383.1 | - | 5 | 9440 | |||
BC025227.1 | - | 8 | 819 | |||
BC025474.1 | - | 9 | 929 | |||
BC120695.1 | - | 24 | 2791 | 40129618 | ||
BC120697.1 | - | 24 | 2791 | 40129620 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
