siRNA Details
Gene Symbol: Hace1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172473.3 | NP_766061.2 | 11 | 1276 | ||
GenBank mRNA | AK029162.1 | - | 1 | 96 | ||
AK042879.1 | - | 11 | 1217 | |||
AK156958.1 | - | 11 | 1218 | |||
BC120695.1 | - | 11 | 1173 | 40129618 | ||
BC120697.1 | - | 11 | 1173 | 40129620 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
