siRNA Details
Gene Symbol: Taar4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008499.1 | NP_001008499.1 | 1 | 646 | ||
GenBank mRNA | BC117729.1 | - | 1 | 645 | ||
BC117730.1 | - | 1 | 635 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Taar4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008499.1 | NP_001008499.1 | 1 | 646 | ||
GenBank mRNA | BC117729.1 | - | 1 | 645 | ||
BC117730.1 | - | 1 | 635 |
Set Membership: |
![]() |