siRNA Details
Gene Symbol: Frmd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177843.1 | NP_001171314.1 | 13 | 1485 | ||
NM_001177844.1 | NP_001171315.1 | 2 | 613 | |||
NM_172475.3 | NP_766063.3 | 13 | 1115 | |||
GenBank mRNA | AK029073.1 | - | 2 | 592 | ||
AK077327.1 | - | 2 | 614 | |||
AK082365.1 | - | 13 | 1061 | |||
AK148414.1 | - | 2 | 612 | |||
AK171532.1 | - | 2 | 541 | |||
BC058672.1 | - | 13 | 1106 | 6830456 | ||
BC082808.1 | - | 13 | 1492 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
