siRNA Details
Gene Symbol: Zfp719 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172482.1 | NP_766070.1 | 4 | 2332 | ||
GenBank mRNA | AK033195.1 | - | 1 | 1711 | ||
AK044684.1 | - | 4 | 2310 | |||
AK083143.1 | - | 4 | 2273 | |||
BC035197.1 | - | 1 | 1292 | |||
BC081548.1 | - | 4 | 2362 |

More Information
Other Species Targeted

Gene Ontology Categories:
