siRNA Details
Gene Symbol: Lrch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081173.1 | NP_001074642.1 | 19 | 2032 | ||
GenBank mRNA | AK122516.1 | - | 7 | 1796 | ||
BC056180.1 | - | 13 | 986 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
