siRNA Details
Gene Symbol: P3h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173379.2 | NP_775555.1 | 4 | 984 | ||
GenBank mRNA | AJ430350.1 | - | 4 | 985 |

More Information
Other Species Targeted

Gene Ontology Categories:
