siRNA Details
Gene Symbol: Tbc1d31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081396.2 | NP_001074865.2 | 14 | 2087 | ||
NM_001167679.1 | NP_001161151.1 | 14 | 2087 | |||
GenBank mRNA | AB185844.1 | - | 14 | 2055 | ||
AK084542.1 | - | 13 | 1851 | |||
AK141805.1 | - | 14 | 1957 | |||
AK154264.1 | - | 14 | 2052 | |||
AK161403.1 | - | 13 | 1904 | |||
AK171024.1 | - | 14 | 2052 | |||
BC050039.1 | - | 14 | 2033 | 30016266 | ||
BC066830.1 | - | 14 | 2019 | 30463017 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
