siRNA Details
Gene Symbol: Hus1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153072.2 | NP_694712.1 | 1 | 652 | ||
GenBank mRNA | AF508546.1 | - | 1 | 565 | ||
AK133107.1 | - | 1 | 654 | |||
BC125607.1 | - | 1 | 645 | 40130122 | ||
BC125609.1 | - | 1 | 645 | 40130124 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
