siRNA Details
Gene Symbol: Ccdc94 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028381.3 | NP_082657.1 | 2 | 93 | ||
GenBank mRNA | AK013533.1 | - | 2 | 107 | ||
AK078481.1 | - | 1 | 1561 | |||
AK168621.1 | - | 2 | 182 | |||
BC031400.1 | - | 2 | 93 | 4952601 |

More Information
Other Species Targeted

Gene Ontology Categories:
