siRNA Details
Gene Symbol: Themis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB509340.1 | - | 2, 3 | 343 | ||
AK040594.1 | - | 2, 3 | 522 | |||
AK041386.1 | - | 2, 3 | 559 | |||
AK041562.1 | - | 2, 3 | 448 | |||
AK041622.1 | - | 2, 3 | 522 | |||
AK088136.1 | - | 2, 3 | 524 | |||
AK089077.1 | - | 2, 3 | 524 | |||
AK138701.1 | - | 2, 3 | 522 | |||
BC125486.1 | - | 2, 3 | 405 | 40130001 | ||
BC125488.1 | - | 2, 3 | 405 | 40130003 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
