siRNA Details
Gene Symbol: Ppp1r36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163103.1 | NP_001156575.1 | 5, 6 | 431 | ||
GenBank mRNA | AK133227.1 | - | 5, 6 | 410 | ||
CK137158.1 | - | 5, 6 | 431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
