siRNA Details
Gene Symbol: Adgrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175642.4 | NP_783573.4 | 28 | 4479 | ||
GenBank mRNA | AK044160.1 | - | 12 | 2238 | ||
AK044248.1 | - | 13 | 1913 | |||
AK081421.1 | - | 7 | 1821 | |||
AY168406.1 | - | 27 | 4218 | |||
BC032251.1 | - | 21 | 2696 | |||
BC062105.1 | - | 12 | 1655 | |||
BC079670.1 | - | 26 | 3498 | |||
BC099951.1 | - | 28 | 4477 | 30098919 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106898, XM_008766929, XM_008766931, XM_008766930, XM_008766928, and others.
Set Membership: |
![]() |
Gene Ontology Categories:
