siRNA Details
Gene Symbol: Gm597 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013750.1 | NP_001013772.1 | 2 | 2138 | ||
GenBank mRNA | AK076972.1 | - | 1 | 57 | ||
BC150996.1 | - | 2 | 2136 | 9087909 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm597 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013750.1 | NP_001013772.1 | 2 | 2138 | ||
GenBank mRNA | AK076972.1 | - | 1 | 57 | ||
BC150996.1 | - | 2 | 2136 | 9087909 |