siRNA Details
Gene Symbol: Kbtbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145958.2 | NP_666070.2 | 4 | 1919 | ||
GenBank mRNA | AK028396.1 | - | 4 | 1865 | ||
AK129374.1 | - | 4 | 1829 | |||
BC022962.1 | - | 1 | 836 | 5368323 | ||
BC023724.1 | - | 1 | 708 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
