siRNA Details
Gene Symbol: March11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177597.5 | NP_808265.2 | 1, 2 | 730 | ||
GenBank mRNA | AK035990.1 | - | 1, 2 | 755 | ||
AK162044.1 | - | 1, 2 | 725 | |||
BC139422.1 | - | 1, 2 | 682 | 8862444 | ||
BC139423.1 | - | 1, 2 | 682 | 8862445 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
