siRNA Details
Gene Symbol: Lrrc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153074.3 | NP_694714.1 | 2 | 922 | ||
GenBank mRNA | AJ422149.1 | - | 1 | 739 | ||
AK080022.1 | - | 2 | 923 | |||
AK171924.1 | - | 2 | 912 | |||
BC137696.1 | - | 2 | 882 | 8860715 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
