siRNA Details
Gene Symbol: Ncoa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172495.5 | NP_766083.3 | 9 | 2177 | ||
GenBank mRNA | AK128935.1 | - | 8 | 2000 | ||
BC042791.1 | - | 257 | ||||
BC076623.1 | - | 7 | 1783 | |||
BC141150.1 | - | 8 | 1878 | 9055762 | ||
BC145459.1 | - | 7 | 1763 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
