siRNA Details
Gene Symbol: Ncoa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172495.5 | NP_766083.3 | 9, 10 | 2260 | ||
GenBank mRNA | AK128935.1 | - | 8, 9 | 2083 | ||
BC042791.1 | - | 340 | ||||
BC076623.1 | - | 7, 8 | 1866 | |||
BC141150.1 | - | 8, 9 | 1961 | 9055762 | ||
BC145459.1 | - | 7, 8 | 1846 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
