siRNA Details
Gene Symbol: Ighv1-54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF045502.1 | - | 2 | 139 | ||
AJ006946.1 | - | 2 | 139 | |||
M15047.1 | - | 2 | 148 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
