siRNA Details
Gene Symbol: 6720489N17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173381.1 | NP_775557.1 | 3 | 1991 | ||
GenBank mRNA | AK033001.1 | - | 3 | 1993 | ||
AK033958.1 | - | 3 | 1975 | |||
BC028252.1 | - | 2158 | ||||
BC053725.1 | - | 3 | 1980 |
