siRNA Details
Gene Symbol: Suox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173733.3 | NP_776094.2 | 4 | 723 | ||
GenBank mRNA | AK149422.1 | - | 4 | 711 | ||
AK154608.1 | - | 4 | 711 | |||
BC027197.1 | - | 3 | 678 | 4160277 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
