siRNA Details
Gene Symbol: Olfr1373 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207227.1 | NP_997110.1 | 1 | 133 | ||
GenBank mRNA | BC106786.1 | - | 1 | 132 | ||
BC106787.1 | - | 1 | 133 | 40041421 | ||
BC106973.1 | - | 1 | 133 | |||
BC119436.1 | - | 1 | 133 | 40041464 |
Gene Symbol: Olfr1371 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207253.1 | NP_997136.1 | 1 | 133 | ||
GenBank mRNA | BC106788.1 | - | 1 | 133 | 40041434 | |
BC106789.1 | - | 1 | 132 |
Gene Symbol: Olfr1381 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146469.2 | NP_666680.2 | 1 | 133 | ||
GenBank mRNA | BC106974.1 | - | 1 | 133 |
Gene Symbol: Olfr1382 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011790.1 | NP_001011790.1 | 1 | 133 | ||
GenBank mRNA | BC119435.1 | - | 1 | 133 | 40041463 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
