siRNA Details
Gene Symbol: Kcnj14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145963.2 | NP_666075.1 | 2 | 774 | ||
GenBank mRNA | AK030711.1 | - | 2 | 784 | ||
AK044725.1 | - | 2 | 755 | |||
AK080732.1 | - | 2 | 763 | |||
BC029692.1 | - | 2 | 714 | 5363132 | ||
BC031753.1 | - | 2 | 774 | 4503027 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
