siRNA Details
Gene Symbol: Tmem87a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110496.1 | NP_001103966.1 | 17 | 1583 | ||
NM_001110497.1 | NP_001103967.1 | 17 | 1583 | |||
NM_173734.3 | NP_776095.2 | 17 | 1586 | |||
GenBank mRNA | AK044591.1 | - | 17 | 1537 | ||
AK086738.1 | - | 17 | 1533 | |||
AK088559.1 | - | 16 | 1494 | |||
BC004047.1 | - | 17 | 1532 | |||
BC024676.1 | - | 7 | 534 | |||
BC027354.1 | - | 17 | 1559 | 4010477 |
