siRNA Details
Gene Symbol: Ap1ar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145964.2 | NP_666076.2 | 5 | 494 | ||
GenBank mRNA | BB611022.1 | - | 5 | 182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
