siRNA Details
Gene Symbol: Tfdp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184706.1 | NP_001171635.1 | 5 | 596 | ||
NM_001184709.1 | NP_001171638.1 | 3 | 220 | |||
NM_001184710.1 | NP_001171639.1 | 2 | 96 | |||
NM_001184711.1 | NP_001171640.1 | 3 | 200 | |||
NM_178667.4 | NP_848782.2 | 6 | 597 | |||
GenBank mRNA | AK029055.1 | - | 5 | 569 | ||
AK033356.1 | - | 3 | 184 | |||
AK053881.1 | - | 5 | 554 | |||
AK079624.1 | - | 6 | 525 | |||
AK158829.1 | - | 2 | 92 | |||
AK162972.1 | - | 5 | 516 | |||
BC092230.1 | - | 3 | 163 | |||
BC141080.1 | - | 4 | 254 | 9055692 | ||
CA980067.1 | - | 3 | 200 | |||
S79780.1 | - | 4 | 279 |
