siRNA Details
Gene Symbol: Plac9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207229.1 | NP_997112.1 | 379 | |||
GenBank mRNA | AK028003.1 | - | 4 | 401 | ||
AK047878.1 | - | 4 | 401 | |||
AK131607.1 | - | 4 | 394 | |||
AK131614.1 | - | 4 | 432 | |||
AK131901.1 | - | 4 | 400 | |||
AK133546.1 | - | 4 | 432 | |||
AK160781.1 | - | 4 | 422 | |||
BC032982.1 | - | 4 | 379 | 1348266 |
Gene Symbol: Plac9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270503.1 | NP_001257432.1 | 4 | 457 | ||
GenBank mRNA | AI391047.1 | - | 4 | 457 |
