siRNA Details
Gene Symbol: Plac9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207229.1 | NP_997112.1 | 182 | |||
GenBank mRNA | AK028003.1 | - | 2 | 204 | ||
AK047878.1 | - | 2 | 204 | |||
AK131614.1 | - | 2 | 235 | |||
AK131901.1 | - | 2 | 203 | |||
AK133546.1 | - | 2 | 235 | |||
AK160781.1 | - | 2 | 225 | |||
BC032982.1 | - | 2 | 182 | 1348266 |
Gene Symbol: Plac9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270503.1 | NP_001257432.1 | 2 | 260 | ||
GenBank mRNA | AI391047.1 | - | 2 | 260 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
