siRNA Details
Gene Symbol: Pcdh9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081377.2 | NP_001074846.1 | 1 | 2503 | ||
NM_001271798.1 | NP_001258727.1 | 2 | 3052 | |||
NM_001271799.1 | NP_001258728.1 | 2 | 2525 | |||
NM_001271800.1 | NP_001258729.1 | 1 | 2503 | |||
GenBank mRNA | AK042546.1 | - | 1 | 1193 | ||
AK147451.1 | - | 2 | 3053 | |||
BC060736.1 | - | 2 | 2485 | |||
BC158114.1 | - | 1 | 2423 | 9007391 | ||
BC158117.1 | - | 1 | 2423 | 9088190 | ||
BC172098.1 | - | 1 | 2421 |
