siRNA Details
Gene Symbol: Trib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144549.4 | NP_653132.1 | 3 | 1207 | ||
GenBank mRNA | AF358866.1 | - | 3 | 790 | ||
AK028626.1 | - | 3 | 1208 | |||
AK044537.1 | - | 3 | 1516 | |||
AK162876.1 | - | 3 | 1208 | |||
BC006800.1 | - | 3 | 791 | 3152842 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
